Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bulk Reagent - ABO & RHD (Rh) Specificities
Products
Brand
Millipore (12)
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
TL-10L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-A from the clone BIRMA-1, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TL-1L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-A from the clone BIRMA-1, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TM-1L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-A,B from the clones ES-15/ES-4, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TP-1L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-D from the clone MS-201, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TL-PACK
Bulk Reagent - ABO & RHD (Rh) Specificities,
CORE Anti-A (clone BIRMA-1) Bulk Reagent plus Diluent Kit
Expand
View Pricing
TM-10L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-A,B from the clones ES-15/ES-4, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TL-50L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-A from the clone BIRMA-1, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TN-1L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-B from the clone LB-2, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TP-10L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-D from the clone MS-201, Human IgM, Blood Grouping Reagent
Expand
View Pricing
TN-PACK
Bulk Reagent - ABO & RHD (Rh) Specificities,
CORE Anti-B (clone LB-2) Bulk Reagent plus Diluent Kit
Expand
View Pricing
TN-10L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-B from the clone LB-2, Murine IgM, Blood Grouping Reagent
Expand
View Pricing
TN-50L-BK
Bulk Reagent - ABO & RHD (Rh) Specificities,
BIOSCOT
™
Anti-B from the clone LB-2, Murine IgM, Blood Grouping Reagent
Expand
View Pricing