Skip to Content
Merck
Anti-Vimentin Antibody serum, Chemicon®

Anti-Vimentin Antibody

Anti-Vimentin Antibody serum, Chemicon®
Synonym(s):
Anti-Vimentin antibody, Mouse monoclonal, Anti-FLJ36605, Anti-VIM

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique