Skip to Content
Merck
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11 clone 22C11, Chemicon®, from mouse

Anti-Beta (β)-Amyloid antibody

Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11 clone 22C11, Chemicon®, from mouse

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2