
8-Bromo-1-octanol

Synonym(s):
Octamethylene bromohydrin
Linear Formula:
Br(CH2)8OH
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing