Skip to Content
Merck
1,6-Hexanedithiol ≥97%, FG

1,6-Hexanedithiol

1,6-Hexanedithiol ≥97%, FG
Synonym(s):
1,6-Dimercaptohexane, DMH, Hexamethylene dimercaptan
Linear Formula:
HS(CH2)6SH
CAS Number:
Molecular Weight:
150.31
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing