Product Name
Anti-LysRS/KARS antibody produced in rabbit
Product Number
SAB5701652
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
Immunohistochemistry | 1:50 - 1:200 |
Immunofluorescence | 1:50 - 1:200 |