Product Name
Monoclonal Anti-LASS3 antibody produced in mouse clone 6C12, purified immunoglobulin
Product Number
SAB1412935
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~47 kDa using MCF-7 lysate. |
Sandwich ELISA | Detection limit for recombinant GST tagged LASS3 is 0.1 ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~34.3 kDa using immunogen protein lysate. |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~42.1 kDa using LASS3 transfected 293T lysate. |