Product Name
ANTI-EPHB3 antibody produced in mouse clone 4E9, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1411943
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | Report Result |
Storage/Stability | Report Result |
Storage Temperature | -20�C |
Species Reactivity | Report Result |
ELISA | Antibody concentration: 1 ug/ml using recombinant protein. |
Sandwich ELISA | Detection limit for recombinant GST tagged EPHB3 is approximately 0.03ng/ml. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1 ug/ml. Specific band of ~36.6 kDa using immunogen protein lysate. |
Western Blotting (Cell Lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~110 kDa using A-431 lysate. |
Western Blotting (Tissue lysate) | Not Verified |
Western Blotting (Transfected lysate) | Not Verified |
Immunohistochemistry | Antibody concentration: 3 ug/ml using human lymph node tissue. |
Immunofluorescence | Not Verified |
Handling | Report Result |