Product Name
Anti-TLR6 (ab1) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
PRS3651
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting | TLR6 antibody can be used for detection of TLR6 by Western blot at 1 to 2 ug/mg/ml. |
ELISA | TLR6 antibody can be used for detection of 10 ng of immunogenic TLR6 peptide by ELISA at 0.01 - 1 ug/ml. |
Immunohistochemistry | Unverified |
Immunofluorescence | TLR6 antibody can be used for immunocytochemistry at 10 ug/ml. |
Immunofluorescence | Unverified |