Product Name
9-cis-Antheraxanthin analytical standard
Product Number
47999
Product Brand
SIAL
CAS Number
Molecular Weight
584.87
Test | Specification |
---|---|
APPEARANCE (COLOR) | Light Yellow to Very Dark Yellow and Light Orange to Very Dark Orange to Very Dark Red and Very Dark Red-Brown |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |