Product Name
trans-2-Phenylcyclopropane-1-carboxylic acid 95%
Product Number
P22354
Product Brand
ALDRICH
CAS Number
939-90-2
Molecular Weight
162.19
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER OR BEADS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 137D OF EDITION I, | |
VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
TITRATION | 94.0%-106.0% (WITH NAOH) |
GAS LIQUID | 94.0% (MINIMUM) |
APPROVED MAY 6, 1997 JSB |