Product Name
4-Methoxycinnamic acid, predominantly trans 99%
Product Number
M13807
Product Brand
ALDRICH
CAS Number
830-09-1
Molecular Weight
178.18
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER AND/OR CHUNKS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 180C OF EDITION I, | |
VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
TITRATION | 98.5%-101.5% (WITH NAOH) |
AND/OR | |
HIGH PRESSURE LIQUID | 98.5% (MINIMUM) |
APPROVED FEBRUARY 21, 1995 JSB |