Product Name
mono-tert-Butyl succinate 97%
Product Number
547352
Product Brand
ALDRICH
CAS Number
15026-17-2
Molecular Weight
174.19
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER, CRYSTALS, CRYSTALLINE POWDER |
AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH NAOH) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED MAY 30, 2008 RJM |