Product Name
2-Chloro-5-iodopyridine 97%
Product Number
498181
Product Brand
ALDRICH
CAS Number
69045-79-0
Molecular Weight
239.44
Test | Specification |
---|---|
APPEARANCE | WHITE TO YELLOW FLAKES OR CRYSTALLINE POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5%(WITH AGNO3 AFTER O2 COMBUSTION) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED FEBRUARY 14, 2006 RJM |