Product Name
(R)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine ≥97.0% (GC)
Product Number
37286
Product Brand
ALDRICH
CAS Number
109838-85-9
Molecular Weight
184.24
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Dark Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.0 % |
SPECIFIC ROTATION (20/D) | -102 ± 5 Degrees |
CONCENTRATION | C=1 IN ETHANOL |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |