Z744205
KDS Stainless Steel Syringe

capacity 200 mL, joint: Swagelok® 1/8, pack of 1 ea
Synonym(s):
liquid injection device, liquid withdrawal device, pump syringe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
42142600
material
stainless steel
Joint
joint: Swagelok® 1/8
packaging
pack of 1 ea
manufacturer/tradename
KD Scientific 780811
capacity
200 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Rugged stainless steel syringes are an ideal solution when the pressures and the force are high, completely eliminating the problem of breaking glass syringes. Both syringe barrel and plungers are #316 stainless steel. An O-ring seal between the top and end of the barrel protects against leakage. Syringes are guaranteed to be leak free for pressures up to 7000 psi
Application
- High pressure applications
- Dispensing fluids
- Withdrawing fluids
Features and Benefits
- Compatable with most syringe pumps
- Eliminate hazards of glass syringe breakage
- Reuseable - fully autoclavable
- Adaptable to Luer Lock or Swagelok® fittings
- Resistance to most chemicals
- Durable construction - Electron beam welded
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service