THS01143
ThalesNano MicroCatCart™ catalyst cartridge system
L × diam. 30 mm × 4 mm, 10% Pt/C
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
Select a Size
Select a Size
About This Item
packaging
pkg of 6 ea
manufacturer/tradename
ThalesNano THS 01143
L × diam.
30 mm × 4 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service