Skip to Content
Merck
All Photos(1)

Key Documents

GF60565091

Copper-beryllium alloy, Cu98Be2

foil, 50 x 50mm, 1.0mm thickness, annealed

Synonym(s):

CU070550

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
31152304
NACRES:
NA.23

form

foil (annealed)

manufacturer/tradename

Goodfellow 605-650-91

size × thickness

50 mm × 50 mm × 1.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service